Anti-ELF1 Rabbit Polyclonal Antibody
AVIVARP36845_P050
- Antibody type:Primary
- Antigen name:E74-like factor 1
- Antigen symbol:ELF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:13709
- Antigen synonyms:Sts1|Elf-1|mElf-1|p70
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: ELF1 (E74-like factor 1 (ets domain transcription factor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: