Order Entry
Spain
ContactUsLinkComponent
Anti-ZNF276 Rabbit Polyclonal Antibody
Anti-ZNF276 Rabbit Polyclonal Antibody
Catalog # AVIVARP36165_T100
Anti-ZNF276 Rabbit Polyclonal Antibody
Catalog # AVIVARP36165_T100

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    zinc finger protein 276
  • Antigen symbol:
    ZNF276
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Epitope:
    middle
  • Cross adsorption:
    none
  • Gene ID:
    92822
  • Antigen synonyms:
    ZNF477|CENPZ|ZADT|ZFP276|CENP-Z
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: ZNF276 (zinc finger protein 276)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: