Anti-ZNF280D Rabbit Polyclonal Antibody
AVIVARP35936_P050
- Antibody type:Primary
- Antigen name:zinc finger protein 280D
- Antigen symbol:ZNF280D
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:54816
- Antigen synonyms:ZNF634|SUHW4
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: ZNF280D (zinc finger protein 280D)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: