Anti-KCTD10 Rabbit Polyclonal Antibody
Catalog # AVIVARP35370_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:potassium channel tetramerization domain containing 10
- Antigen symbol:KCTD10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:83892
- Antigen synonyms:ULRO61|hBACURD3|MSTP028|BTBD28
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: KCTD10 (potassium channel tetramerization domain containing 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: