Anti-ZNF202 Rabbit Polyclonal Antibody
Catalog # AVIVARP33549_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:zinc finger protein 202
- Antigen symbol:ZNF202
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:7753
- Antigen synonyms:ZSCAN42|ZKSCAN10
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: ZNF202 (zinc finger protein 202)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: