Order Entry
Spain
ContactUsLinkComponent
Anti-CCL11 Rabbit Polyclonal Antibody
Anti-CCL11 Rabbit Polyclonal Antibody
  AVIVARP30865_P050
Anti-CCL11 Rabbit Polyclonal Antibody
  AVIVARP30865_P050

 

  • Antibody type:
    Primary
  • Antigen name:
    chemokine (C-C motif) ligand 11
  • Antigen symbol:
    CCL11
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Cross adsorption:
    none
  • Gene ID:
    20292
  • Antigen synonyms:
    eotaxin|Scya11
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: CCL11 (chemokine (C-C motif) ligand 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: