Order Entry
Spain
ContactUsLinkComponent
Anti-CCL7 Rabbit Polyclonal Antibody
Anti-CCL7 Rabbit Polyclonal Antibody
Catalog # AVIVARP30804_P050
Anti-CCL7 Rabbit Polyclonal Antibody
Catalog # AVIVARP30804_P050

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    chemokine (C-C motif) ligand 7
  • Antigen symbol:
    CCL7
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Cross adsorption:
    none
  • Gene ID:
    6354
  • Antigen synonyms:
    MCP3|NC28|SCYA7|SCYA6|MCP-3|MARC|FIC
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: CCL7 (chemokine (C-C motif) ligand 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human