Anti-HTR2B Rabbit Polyclonal Antibody
Catalog # AVIVARP30716_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:5-hydroxytryptamine (serotonin) receptor 2B
- Antigen symbol:HTR2B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:3357
- Antigen synonyms:5-HT2B|5-HT(2B)
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: HTR2B (5-hydroxytryptamine (serotonin) receptor 2B, G protein-coupled)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: