Anti-FADD Rabbit Polyclonal Antibody
Catalog # AVIVARP30294_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Fas (TNFRSF6)-associated via death domain
- Antigen symbol:FADD
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Mouse
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:14082
- Antigen synonyms:Mort1/FADD
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: FADD (Fas (TNFRSF6)-associated via death domain)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Mouse