Anti-CX3CL1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-0811R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Chemokine (C-X3-C Motif) Ligand 1
- Antigen symbol:CX3CL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:CXC3|fractalkine|NTN|CXC3C|SCYD1|ABCD-3|C3Xkine|NTT|neurotactin
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: CX3CL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: