Order Entry
Spain
ContactUsLinkComponent
Disposable columns, DISPOSAFLEX®, KIMBLE®
Disposable columns, DISPOSAFLEX®, KIMBLE®
Catalog # SCERSP420160-0000
Supplier:  DWK Life Sciences
Disposable columns, DISPOSAFLEX®, KIMBLE®
Catalog # SCERSP420160-0000
Supplier:  DWK Life Sciences

Specifications

  • Ø int.:
    8,00 mm
  • Capacity:
    8 ml
  • Description:
    DISPOSAFLEX® column set, unassembled
  • Length:
    200 mm
  • Pk:
    100

Specifications

About this item

This is a multi purpose column designed to satisfy a wide range of analytical separations using aqueous buffers and typically used for the extraction of the cyclic nucleotides cAMP and cGMP from sample preparations using AG1-X8.

  • Eliminates the possibility of cross contamination

The lower column fitting with a Luer outlet is molded PP which is designed to secure hold a porous PE bed support or a glass wool plug.