



Sodium chlorite, anhydrous ≥79%, flakes, J.T.Baker®
V044-07
: Avantor
:
- Pack type:Glass bottle, amber, wide mouth
- Pk:500 g
- Seq:3645
- For Laboratory,Research,or Manufacturing Use:= 79 %
- Assay (NaClO₂):≥79 %
- Appearance:Passes Test
:
:90.44 g/mol
:13,08672
:112 °C (1013 hPa)
:250…257 °C
:1.28 g/cm³ (20 °C)
:Ambient
:231-836-6
:1496
:5.1,II
:GHS03
:GHS06
:GHS05
:GHS09
:7758-19-2
:ClNaO₂
:MFCD00003478