Order Entry
Spain
ContactUsLinkComponent
Capillary tubes
Capillary tubes
Catalog # SARS19.447.001
Supplier:  SARSTEDT
undefined
Capillary tubes
Catalog # SARS19.447.001
Supplier:  SARSTEDT
Specifications
  • Ø:
    1,50 mm
  • Volume:
    20 µl
  • Description:
    Capillary Tubes
  • Pk:
    1.000
Specifications
About this item
Glass, transparent.
  • End-to-end capillaries
  • Prepared with potassium EDTA