Inspection mirror
Catalog # SIMELD3025
Supplier: Industrial Physics
Specifications
- Description:Robust telescopic mirror, glass/metal, 100 g
- Length:254/375 (in/out)
- Seq:02
- Pk:1
- Ø:56 mm
Specifications
About this item
Ideal for visual inspection of hard-to-reach spots. Spots that are difficult to inspect are even more difficult to blast or coat.
- With a simple inspection mirror a visual inspection behind stiffeners, under- or on top of I-beams or other awkward places is easy to perform
- Made of clear glass that can be positioned by setting the double all angle ball joints for for 360° viewing
- Ball joints are manufactured from spring steel to withstand stresses of constant repositioning and hold the mirror in any desired position