Order Entry
Spain
ContactUsLinkComponent
Perfluoropolyether oil for rotary vacuum pumps, Fomblin® YVAC
Perfluoropolyether oil for rotary vacuum pumps, Fomblin® YVAC
  EDWAH11301019
Perfluoropolyether oil for rotary vacuum pumps, Fomblin® YVAC
  EDWAH11301019
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Description:
    Rotary vacuum pump oil, perfluoropolyether, 532 ml
  • Seq:
    01
  • Pk:
    1 kg

 

 

Fomblin® perfluoropolyether oil for use in oil sealed rotary vacuum pumps and are practically a direct replacement for mineral oil in terms of viscosity and vapour pressure. Also it contains Drynert fluid which contains anti-rust and anti-wear additives soluble in Fomblin®. These additives cover metallic surfaces with a protective, corrosion resistant film.

It is widely used for semiconductors, chemical pumping, oxygen pumping, radioactive and metallisation.