Chemical resistant overalls, Tyvek® 800 J, model TJ 198 Ta
Supplier: DuPont
Certificates
About this item
This robust yet lightweight garment benefits from the use of a specific 'impervious technology' to achieve an effective barrier against many low-concentrated water-based inorganic chemicals (even under pressure) and small-sized hazardous particulates, as well as oil repellences.
- Anti-static treatment on inside
- Robust yet lightweight (<300 g)
- Hood, Tyvek® zip and self-adhesive zip flap for enhanced protection
- Self-adhesive chin flap for tight seal of suit to mask
- Elasticated face, wrists and ankles as well as glued-in waist elastic for good fit
- Thumb loops to prevent sleeves from riding up
- Stitched and over-taped seams (orange) for protection and strength
Ideal for use in very humid applications requiring chemical, liquid protection and/or oil repellences. Typical activities include industrial cleaning, work at petrochemical installations, in sewers and maintenance operations.
Weight: 59 g/m²
Colour: White (orange seams)
Certifications: EN 1073-2, EN 1149-5, EN 14126, Type 3-B (Additional taping of cuffs, ankles, hood and zip flap required to achieve Type 3 liquid tightness); Type 4-B/5-B/6-B; Cat. III
Packaging: 25 per box, individually packed.
Specifications
- Catalog No:
- DUPOTJ198TWHXL0025
- DUPOTJ198TWH3X0025
- DUPOTJ198TWHLG0025
- DUPOTJ198TWH4X0025
- DUPOTJ198TWHSM0025
- DUPOTJ198TWH5X0025
- DUPOTJ198TWH6X0025
- DUPOTJ198TWHMD0025
- DUPOTJ198TWH2X0025
- DUPOTJ198TWH7X0025
- Model:
- 800 J, TJ 198 Ta
- 800 J, TJ 198 Ta
- 800 J, TJ 198 Ta
- 800 J, TJ 198 Ta
- 800 J, TJ 198 Ta
- 800 J, TJ 198 Ta
- 800 J, TJ 198 Ta
- 800 J, TJ 198 Ta
- 800 J, TJ 198 Ta
- 800 J, TJ 198 Ta
- Anti-static:
- +
- +
- +
- +
- +
- +
- +
- +
- +
- +
- Garment application:
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Weight:
- 59 g/m²
- 59 g/m²
- 59 g/m²
- 59 g/m²
- 59 g/m²
- 59 g/m²
- 59 g/m²
- 59 g/m²
- 59 g/m²
- 59 g/m²
Specifications
Frequently Bought Together