Order Entry
Spain
ContactUsLinkComponent
Media preparator, ProfiClave
Media preparator, ProfiClave
  710-1670
 :  BioTool
undefined
Media preparator, ProfiClave
  710-1670
 :  BioTool
  • Description:
    ProfiClave PC20B
  • Pk:
    1
The media preparators ProfiClave PC10B/20B prepare up to 10/15 L of plain or complex vitamin/blood added agar formulations easier and faster than ever. Equipped with intuitive software, accessible via a convenient touch screen, training on how to use the media preparator is short and easy. ProfiClave PC20 prepares between 1 to 15 L, ProfiClave PC10 prepares between 0,5 to 10 L of agar, stirred and temperature controlled. Vessel access is simple, no bolts or tools required. Enhanced user programming capabilities (3 * 20 programs), operating parameter print-out (integrated in front panel), external control functions (front panel connection), optimal stirring control and full metal housing makes the ProfiClave one of the best media preparators in its class. It accurately maintains the agar in any stage of the single or double cycle process at an even temperature for hours. Water jacket design enables even heating within the vessel. Stirrer with variable speeds and enhanced magnetic coupling without seals or shafts.
  • Easy operation software guides smooth transition and a user friendly touch screen
  • Dish stacks of required height can be freely selected (1 to 20 dishes/stack)
  • Media smoothing allows optimisation of the filling temperature
  • System utilises bevelled edge/automation plates
  • High sterility at pouring area due to well positioned UV light (with safety switch)
  • Smother agar plates using dual head pump
  • Repels of agar prevented by an anti drop mechanism
  • Easy cleaning process due to ergonomic design and easy access