Multi-gas detector, GasAlert Max XT II
The rugged GasAlert Max XT II reliably monitors up to four gas hazards and combines straightforward one-button field operation with an integrated sampling pump.
- Compatible with BW MicroDock II automatic test and calibration station
- Powered by a rechargeable battery pack
- GasAlert Max XT II comes with standard event logging and datalogging (accessory connectivity kit required)
Tamper-proof, user-adjustable options enable the instrument to be customised to suit your application.
: Supplied with detector complete with sensors, stainless steel alligator clip and concussion-proof housing, rechargeable lithium polymer battery, charging adaptor, calibration hose with quick connect (1 m), sampling hose with particulate filter and quick connect (3 m), reversible screwdriver, five spare pump filters, manual and multi-language CD manual.
- Catalog No:
- BWTEXT-0W0M-Y-EU
- 151-0239
- BWTEXT-XW00-Y-EU
- BWTEXT-X00M-Y-EU
- BWTEXT-0W00-Y-EU
- BWTEXT-X000-Y-EU
- BWTEXT-XW0M-Y-EU
- Model:
- GasAlert Max XT II
- GasAlert Max XT II
- GasAlert Max XT II
- GasAlert Max XT II
- GasAlert Max XT II
- GasAlert Max XT II
- GasAlert Max XT II
- Alarm:
- Visual, vibrating, audible (95 dB)
- Visual, vibrating, audible (95 dB)
- Visual, vibrating, audible (95 dB)
- Visual, vibrating, audible (95 dB)
- Visual, vibrating, audible (95 dB)
- Visual, vibrating, audible (95 dB)
- Visual, vibrating, audible (95 dB)
- Battery life:
- 10 hours at −20 °C, 13 hours at 0 °C
- 10 hours at −20 °C, 13 hours at 0 °C
- 10 hours at −20 °C, 13 hours at 0 °C
- 10 hours at −20 °C, 13 hours at 0 °C
- 10 hours at −20 °C, 13 hours at 0 °C
- 10 hours at −20 °C, 13 hours at 0 °C
- 10 hours at −20 °C, 13 hours at 0 °C
- W×D×H:
- 7.0×5.2×13.1 cm
- 7.0×5.2×13.1 cm
- 7.0×5.2×13.1 cm
- 7.0×5.2×13.1 cm
- 7.0×5.2×13.1 cm
- 7.0×5.2×13.1 cm
- 7.0×5.2×13.1 cm
- Weight:
- 316 g
- 316 g
- 316 g
- 316 g
- 316 g
- 316 g
- 316 g
Frequently Bought Together