Centrifuge tubes, with screw caps, EUROTUBO®
: DELTALAB
Made of PP, translucent or PS, clear, with green HDPE screw cap.
- PP: autoclavable
- PP: Withstand forces up to 15000×g
- PS: Withstand forces up to 7500×g
The external skirt allows the tubes to remain free-standing. Supplied screwed. The sterile model (ethylene oxide) is supplied individually in flow-pack bag, with indication of batch, expiration date, etc. according to the guidelines for sterile products.
: For autoclaving, the cap should be loose on the thread and not tightly fitted.
Frequently Bought Together