Chromatography paper
Supplier: VARIAN
Certificates
About this item
Chromatography paper is used in thin layer chromatography applications such as evaluating radioisotope purity, QC testing.
- The porous paper is made of glass microfibers impregnated with silica gel
- No interference from organic binders
- Separates lipids and other non-polar compounds
- Can easily be cut to convenient testing sizes, and can be imprinted
Ordering information: Offers two kinds of paper such as SA (contains sodium salt) and SG (contains potassium salt).
Frequently Bought Together