Order Entry
Spain
ContactUsLinkComponent
Tubing cutter for fused silica tubing, Upchurch Scientific®
Tubing cutter for fused silica tubing, Upchurch Scientific®
Catalog # UPCHFS-315
Tubing cutter for fused silica tubing, Upchurch Scientific®
Catalog # UPCHFS-315
Specifications
  • Description:
    Fused silica tubing cutter, Shortix™, for tubing O.Ø 350 - 780 µm and I.Ø 100 - 350 µm
  • Seq:
    02
  • Pk:
    1
Specifications
About this item
Precision cutter for fused silica tubing ensures clean, trouble free cutting of fused silica tubing.
It also includes a built-in magnifying glass to examine the cut tubing ends.