Order Entry
Spain
ContactUsLinkComponent
Orthoboric acid 6 g/l in aqueous solution
Orthoboric acid 6 g/l in aqueous solution
Catalog # PRLS88973.360
Supplier:  PROLABO CMP
CAS Number:  
undefined
Orthoboric acid 6 g/l in aqueous solution
Catalog # PRLS88973.360
Supplier:  PROLABO CMP
CAS Number:  

Specifications

  • Pk:
    5 L
  • Seq:
    5190

Specifications

About this item

 :
 :61.83 g/mol
Storage Temperature:Ambient
EINECS:233-139-2
CAS Number:10043-35-3
Formula:B(OH)₃
MDL Number:MFCD00011337