Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Spain
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
ortho-Boric acid
Orthoboric acid in aqueous solution
Orthoboric acid in aqueous solution
Orthoboric acid in aqueous solution
PRLS88143.290
:
PROLABO CMP
:
Orthoboric acid in aqueous solution
PRLS88143.290
:
PROLABO CMP
:
Pack type:
Glass bottle
Pk:
1 L
Seq:
5037
:
:
61.83 g/mol
:
Ambient
:
233-139-2
:
10043-35-3
:
B(OH)₃
:
MFCD00011337