Order Entry
Spain
ContactUsLinkComponent
Orthoboric acid 20 g/l in aqueous solution, with indicator
Orthoboric acid 20 g/l in aqueous solution, with indicator
  PRLS85767.360
 :  
undefined
Orthoboric acid 20 g/l in aqueous solution, with indicator
  PRLS85767.360
 :  

 

  • Pack type:
    Plastic container
  • Pk:
    5 L
  • Seq:
    5190

 

 

 :
 :61.83 g/mol
 :Ambient
 :233-139-2
 :10043-35-3
 :B(OH)₃
 :MFCD00011337