Order Entry
Spain
ContactUsLinkComponent
Rhodamine B ≥98.0% (by titrimetric analysis) ion association reagent for photometric and fluorimetric analysis

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Green to Dark green powder to crystal
  • Purity (Nonaqueous Titration):
    min. 98.0 % (calcd.on dried substance)
  • Lambda max.:
    552.0 to 556.0 nm (H2O)
  • Absorbance (E1%1cm):
    min. 2,000 (H2O, 552.0 to 556.0 nm)
  • Solubility in Water:
    almost transparency

Specification Test Results

About this item

Synonyms: 9-(2-Carboxyphenyl)-3,6-bis(diethylamino)xanthylium chloride, [9-(2-Carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium chloride
 :
 :479.02 g/mol
Merck Index:14,08183
Melting Pt:199…201 °C
Density:1,31 g/cm³ (20 °C)
EINECS:201-383-9
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:81-88-9
Formula:C₂₈H₃₁ClN₂O₃
MDL Number:MFCD00011931