
1-(4-Chlorophenyl)cyclopropanecarboxylic acid 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:72934-37-3
:
Formula:C₁₀H₉ClO₂
MDL Number:MFCD00001289
:196.64 g/mol
Melting Pt:152…155 °C
EINECS:277-105-5
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:152 to 157 °C
- Titration with NaOH:≥98.5 %
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together