1-(3-Chlorophenyl)piperazine hydrochloride
Catalog # PROOMM0421.09
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:196.68 g/mol (anhy)
EINECS:265-718-0
CAS Number:65369-76-8
Formula:C₁₀H₁₃ClN₂·xHCl
MDL Number:MFCD00039032