Phenylacetaldehyde 40 - 55% in diethyl phthalate
TCIAP0119-100G
: TCI
:
- Pack type:Glass bottle
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
- Appearance:White to Yellow to Green clear liquid
- Concentration (Oxime Formation):40.0 to 55.0 w/w%
: Phenyl acetic aldehyde
:
:120.15 g/mol
:204-574-5
:GHS07
:Refrigerator
:122-78-1
:C₈H₈O
:MFCD00006993
:195
:14,07265