Cellulose chromatography papers
Supplier: MACHEREY-NAGEL
Certificates
About this item
Paper chromatography is the chromatographic technique separation due to partition of the analytes between special paper grades and the mobile phase, which penetrates the paper by capillary action ascending, descending and circular techniques are possible.
Specifications
- Catalog No:
- MANA817004A
- MANA817003A
- Grade:
- MN 261
- MN 260
- Thickness:
- 0,18 mm
- 0,20 mm
- Flow rate:
- 90 - 100 mm / 30 min
- 120 - 130 mm / 30 min
- Weight:
- 90 g/m²
- 90 g/m²