Disposable weighing boats, anti-static, Sterilin®
Supplier: STERILIN
Certificates
About this item
PS, diamond shaped.
- Avoids the retention of powders by static electricity
- Ideal for use with fine powders e.g. talcum powder
Raw material modified to significantly reduce the charge relaxation time in comparison to standard weigh boats. Majority of electrostatic charge dissipates within seconds when earthed on the balance pan.
Frequently Bought Together