Order Entry
Spain
ContactUsLinkComponent
VWR® YPD, Media, Liquid, for Biotechnology
VWR® YPD, Media, Liquid, for Biotechnology
Catalog # 1B1493-1L
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® YPD, Media, Liquid, for Biotechnology
Catalog # 1B1493-1L
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Description:
    YPD media, liquid

Specifications

About this item

A complete medium for the propagation and maintenance of yeast. The composition of YPD is (w/v) 1% yeast extract, 2% peptone, and 2% D-glucose.

pH (25 °C): 6,14 to 7,14