Ion exchange HPLC columns, BioBasic™ AX and SCX
Proveedor: Thermo Fisher Scientific
Certificados
Acerca de este artículo
BioBasic™ AX LC columns optimized for the separation of proteins, peptides, other anionic species and highly polar molecules.
- Low hydrophobicity
- Use for normal phase and hydrophilic interaction chromatography (HILIC)
- Weak anion exchange phase for multiple charged species
Especificaciones
- Catalog No:
- HYPE73205-251030
- HYPE73205-151030
- HYPE73205-152130
- HYPE73205-101030
- HYPE73105-153030
- HYPE73205-052130
- HYPE73105-151030
- HYPE73205-154630
- HYPE73205-014001
- HYPE73105-054630
- HYPE73105-254630
- HYPE73205-102130
- HYPE73105-051030
- HYPE73105-152130
- HYPE73205-104630
- HYPE73205-053030
- HYPE73205-054630
- HYPE73105-052130
- HYPE73105-101030
- HYPE73105-154630
- HYPE73105-053030
- HYPE73105-104630
- HYPE73105-012101
- HYPE73205-254630
- HYPE73105-102130
- Carbon load:
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- 3%
- pH range:
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- USP code:
- L52
- L52
- L52
- L52
- L52
- L52
- L52
- L52
- L52
- L52
- L52
- L52
Especificaciones
Frequently Bought Together