Fittings, LuerTight™, Upchurch Scientific®
Supplier: UPCHURCH SCIENTIFIC
Certificates
About this item
LuerTight™ fittings are designed for connecting fluoropolymer tubing without barbs or nuts. By integrating ferrules into the Luer bodies, these fitting reliably hold tubing in place while giving the convenience of a Luer connection.
- An inline set of fittings provides a quick and easy disconnection option
- The bodies of these are manufactured from PP and the ferrules are made of ETFE
Frequently Bought Together