Order Entry
Spain
ContactUsLinkComponent
Acetic acid 27%
Acetic acid 27%
Catalog # PRLS85558.360
Supplier:  PROLABO CMP
CAS Number:  
undefined
Acetic acid 27%
Catalog # PRLS85558.360
Supplier:  PROLABO CMP
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 L
  • Seq:
    5190

Specifications

About this item

 :
 :60.05 g/mol
Storage Temperature:Ambient
EINECS:200-580-7
UN:2790
ADR:8,III
 :GHS05
CAS Number:64-19-7
Formula:H₃CCOOH
MDL Number:MFCD00036152