Order Entry
Spain
ContactUsLinkComponent
Phenolphthalein 0.8% (w/w) in ethanol
Phenolphthalein 0.8% (w/w) in ethanol
Catalog # PRLS88653.290
Supplier:  PROLABO CMP
CAS Number:  
undefined
Phenolphthalein 0.8% (w/w) in ethanol
Catalog # PRLS88653.290
Supplier:  PROLABO CMP
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :318.33 g/mol
Storage Temperature:Ambient
EINECS:201-004-7
UN:1170
ADR:3,II
 :GHS02
CAS Number:77-09-8
Formula:C₂₀H₁₄O₄
MDL Number:MFCD00005913