Order Entry
Spain
ContactUsLinkComponent
Trichloroacetic acid (TCA) 150 g/l in aqueous solution
 
 
 
Trichloroacetic acid (TCA) 150 g/l in aqueous solution
  PRLS87657.360
 :  
undefined
 
 
 
Trichloroacetic acid (TCA) 150 g/l in aqueous solution
  PRLS87657.360
 :  

 

  • Pk:
    5 L
  • Seq:
    5190

 

 

 :
 :163.39 g/mol
 :196 °C (1013 hPa)
 :54…56 °C
 :≤1.07 g/cm³ (20 °C)
 :Ambient
 :200-927-2
 :2564
 :8,II
 :GHS05
 :GHS07
 :GHS09
 :76-03-9
 :Cl₃CCOOH
 :MFCD00004177