Order Entry
Spain
ContactUsLinkComponent
Potassium hydroxide 130 g/l in aqueous solution
Corrosive
Potassium hydroxide 130 g/l in aqueous solution
Catalog # PRLSMC306234.2000
Supplier:  PROLABO CMP
CAS Number:  
Corrosive
Potassium hydroxide 130 g/l in aqueous solution
Catalog # PRLSMC306234.2000
Supplier:  PROLABO CMP
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2 L
  • Seq:
    5086

Specifications

About this item

 :
 :56.11 g/mol
Storage Temperature:Ambient
EINECS:215-181-3
UN:1814
ADR:8,II
 :GHS05
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553