Order Entry
Spain
ContactUsLinkComponent
Silver nitrate 0.005 mol/l (0.005 N) in isopropanol (IPA)
Flammable
Irritant
Silver nitrate 0.005 mol/l (0.005 N) in isopropanol (IPA)
Catalog # PRLS88016.360
Supplier:  PROLABO CMP
CAS Number:  
undefined
Flammable
Irritant
Silver nitrate 0.005 mol/l (0.005 N) in isopropanol (IPA)
Catalog # PRLS88016.360
Supplier:  PROLABO CMP
CAS Number:  

Specifications

  • Pk:
    5 L
  • Seq:
    5190

Specifications

About this item

 :
 :169.87 g/mol
Storage Temperature:Ambient
EINECS:231-853-9
UN:1993
ADR:3,II
 :GHS02
 :GHS07
CAS Number:7761-88-8
Formula:AgNO₃
MDL Number:MFCD00003414