2-Methyl-4,5-diphenyloxazole
Supplier: TCI
Certificates
About this item
CAS Number:14224-99-8
:
Formula:C₁₆H₁₃NO
MDL Number:MFCD00005307
Boiling Pt:212°C
Specification Test Results
- Appearance:Light yellow to Yellow to Orange clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together