Order Entry
Spain
Orders LinkContactUsLinkComponent
LaserTab laser printable labels
LaserTab laser printable labels
 :   Brady
LaserTab laser printable labels
 :  Brady
LaserTab laser printable labels
LaserTab laser printable labels
Laboratory labels are compatible with office laser printers. The high-performance lab polyester labels feature a low profile to prevent vials from jamming in centrifuges and racks and they can be used in liquid nitrogen to –196 °C (–321 °F) and in autoclaves up to 121 °C (250 °F).
Product Family Options
  • Description
    Model
    Material
    Adhesive
    Printing method
    Colour
    Dimensions
    Packed
  • Printer labels
    LAT-31-759-1
    Paper
    Permanent acrylic
    Laser
    White
    50,0×22,86 mm
    1020 labels/pack
    1 * 1.020 UN
    1 * 1.020 UN - 124,00 €
    124,00 €
    1 * 1.020 UN
       124,00 €
  • Printer labels
    LAT-13-707-2.5
    Polyester
    Permanent acrylic
    Laser
    White
    47,63×21,16 mm
    2508 labels/pack
    1 * 2.508 UN
    1 * 2.508 UN - 548,00 €
    548,00 €
    1 * 2.508 UN
       548,00 €
  • Printer labels, cloth
    LAT-61-799-2.5
    Nylon
    Permanent acrylic
    Laser
    White
    Ø: 12,70 mm
    2500 labels/pack
    1 * 2.500 UN
    1 * 2.500 UN - 172,00 €
    172,00 €
    1 * 2.500 UN
       172,00 €