VWR® ARISTAR®, Multi-Element Quality Control Standard 100 mg/L, 9 Elements in Nitric Acid 5% for ICP-MS
Catalog # 88175.230
Supplier: VWR Chemicals
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Plastic bottle
- Pk:250 mL
- Description:Multi-element quality control standard 100 mg/L {Be; I; Bi; Li; Cr; Mg; Co; Pb; U} in nitric acid 5% ARISTAR® for ICP-MS
Specifications
About this item
9 Elements 100 mg/L: Be, In, Bi, Li, Cr, Mg, Co, Pb, U
Certifications: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Delivery: Supplied with certificate of analysis.