External national pipe thread adapters, Upchurch Scientific®
Supplier: UPCHURCH SCIENTIFIC
Certificates
About this item
These PEEK adapters make connections to female ¹/₈" and ¹/₄" National Pipe Thread (NPT) ports.
- Chemically resistant
- Pressure rating up to 500 psi (34 bar)
Frequently Bought Together