PEEK frits, Upchurch Scientific®
Supplier: UPCHURCH SCIENTIFIC
Certificates
About this item
Exceptionally uniform porosity which ensures longer filtration life and consistent frit-to-frit swept volumes. These frits are biocompatible and inert to most solvents, making them ideal for bioanalytical applications. The disc rings, made of PCTFE, are slightly thicker than the frit disc, providing enhanced sealing and excellent chemical resistance.
- Inert, biocompatible and metal-free
- PCTFE surrounded PEEK frits can be used up to 80 °C, others up to 100 °C
Frequently Bought Together