Order Entry
Spain
ContactUsLinkComponent
Capillary columns, CP-Wax 51 for amines
Capillary columns, CP-Wax 51 for amines
Catalog # VARICP7405
Supplier:  VARIAN
undefined
Capillary columns, CP-Wax 51 for amines
Catalog # VARICP7405
Supplier:  VARIAN

Specifications

  • Column Format:
    Capillary
  • Film thickness:
    0,20 µm
  • Ø int.:
    0,25 mm
  • Description:
    Temperature limits 60 to 260/275 °C, 7 in cage
  • Length:
    25 m
  • Phase:
    CP-Wax 51 for amines
  • Pk:
    1

Specifications

About this item

CP-Wax 51 for amines is designed for the analysis of C4-C10 amines, diamines and aromatic amines.

  • Symmetrical peaks due to basic reagent deactivation
Certifications: Conforms to EPA method 607.