Order Entry
Spain
ContactUsLinkComponent
Pesticide reference standard, 1,3,5-Trichlorobenzene 1,000 µg/ml in cyclohexane
Pesticide reference standard, 1,3,5-Trichlorobenzene 1,000 µg/ml in cyclohexane
Catalog # PROONE5712
CAS Number:  
Pesticide reference standard, 1,3,5-Trichlorobenzene 1,000 µg/ml in cyclohexane
Catalog # PROONE5712
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1,5 mL
  • CAS No.:
    108-70-3
  • Description:
    1,3,5-Trichlorobenzene 1,000 µg/ml in cyclohexane
  • Formula:
    C₆H₃Cl₃
  • Description:
    1,3,5-Trichlorobenzene

Specifications

About this item