Order Entry
Spain
ContactUsLinkComponent
1,2-Dichlorobenzene-[D4] 2000 µg/ml in methanol, Dr Ehrenstorfer
1,2-Dichlorobenzene-[D4] 2000 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERYA12370100ME
Supplier:  EHRENSTORFER
CAS Number:  
1,2-Dichlorobenzene-[D4] 2000 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERYA12370100ME
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    2199-69-1
  • Description:
    1,2-Dichlorobenzene-[D4] 2,000 µg/ml in methanol
  • Formula:
    C₆₂H₄Cl₂
  • Description:
    1,2-Dichlorobenzene D4

Specifications

About this item