Order Entry
Spain
ContactUsLinkComponent
Perchloric acid, BAKER ANALYZED® ACS, J.T.Baker®
Perchloric acid, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Perchloric acid, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    60.0 - 62.0 %
  • ACS - Assay (HClO₄):
    60.0 - 62.0 %
  • ACS - Color (APHA):
    ≤10
  • ACS - Residue after Ignition:
    ≤0.003 %
  • ACS - Chloride (Cl):
    ≤0.001 %
  • ACS - Nitrogen Compounds (as N):
    ≤0.001 %
  • ACS - Sulfate (SO₄):
    ≤0.001 %
  • ACS - Silicate and Phosphate (as SiO₂):
    ≤5 ppm
  • ACS - Heavy Metals (as Pb):
    ≤1 ppm
  • Trace Impurities - Iron (Fe):
    ≤1 ppm

Specification Test Results

Related Information

Frequently Bought Together