Order Entry
Spain
Orders LinkContactUsLinkComponent
Bromophenol blue ACS

Bromophenol blue ACS

Supplier:   G-Biosciences
Bromophenol blue ACS
Supplier:  G-Biosciences
Certificates
Bromophenol blue ACS

3 Options Available Below

Product Details & Documents
Synonyms:  Bromphenol Blue Sultone Form, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein
  • CAS Number:115-39-9
  • Formula:C₁₉H₁₀Br₄O₅S
  • MDL Number:MFCD00005875
  • Molecular Weight:669.97 g/mol
  • Merck Index:13,01429
  • Melting Pt:273 °C
  • Density:1 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:204-086-2
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 90,40 €
    90,40 €
    1 * 25 g
       90,40 €

  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - 168,00 €
    168,00 €
    1 * 50 g
       168,00 €

  • Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 1 490,00 €
    1 490,00 €
    1 * 500 g
       1 490,00 €