Tetradecanophenone ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:4497-05-6
:
Formula:C₂₀H₃₂O
MDL Number:MFCD00008985
Boiling Pt:206°C
EINECS:224-790-3
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:51.0 to 54.0 deg-C
- Solubility in hot Methanol:almost transparency
Specification Test Results
Frequently Bought Together