0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Spain
ContactUsLinkComponent
Standards
Organic Standards
Organic reference standard, Theophyllin 10 µg/ml in methanol
Organic reference standard, Theophyllin 10 µg/ml in methanol
PROONE9329
:
LGC Standards PROMOCHEM
:
Organic reference standard, Theophyllin 10 µg/ml in methanol
PROONE9329
:
LGC Standards PROMOCHEM
:
Pack type:
Glass bottle
Pk:
10 mL
CAS No.:
58-55-9
Description:
Theophyllin 10 µg/ml in methanol
Formula:
C₇H₈N₄O₂
Description:
Theophyllin